Learn More
GBA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 171641AP
Catalog No. 50-173-1411
Add to cart
Description
Description
Non-lysosomal glucosylceramidase that catalyzes the conversion of glucosylceramide (GlcCer) to free glucose and ceramide. Involved in sphingomyelin generation and prevention of glycolipid accumulation. May also catalyze the hydrolysis of bile acid 3-O-glucosides, however, the relevance of such activity is unclear in vivo. Plays a role in central nevous system development. Required for proper formation of motor neuron axons.
Specifications
Specifications
GBA2
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5M868, Q69ZF3, Q9HCG7
AD035, Beta glucocerebrosidase 2, Beta glucosidase 2, GBA2, Glucosylceramidase 2, KIAA1605, NLGase
Rabbit
IgG
150 μL
-20°C
Primary
230101, 298399, 57704
ELISA
Unconjugated
GBA2
Liquid
GBA2
GBA2 Fusion Protein Ag10606
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat