Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GBA3 Mouse anti-Human, Clone: OTI5G3, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502605

Catalog No. 50-167-3644


Add to cart

Description

Description

GBA3, or cytosolic beta-glucosidase (EC 3.2.1.21), is a predomitly liver enzyme that efficiently hydrolyzes beta-D-glucoside and beta-D-galactoside, but not any known physiologic beta-glycoside, suggesting that it may be involved in detoxification of plant glycosides. GBA3 also has significant neutral glycosylceramidase activity (EC 3.2.1.62), suggesting that it may be involved in a nonlysosomal catabolic pathway of glucosylceramide metabolism.
Specifications

Specifications

GBA3
Monoclonal
0.72 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9H227
CBG, CBGL1, GLUC, KLRP
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57733
Flow Cytometry, Western Blot
OTI5G3
Unconjugated
GBA3
Liquid
GBA3
Human recombit protein fragment corresponding to amino acids 1-150 and 370-469 of human GBA3 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit