Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GBE1 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI1D11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270810AF647

Catalog No. N270810A647


Add to cart

Description

Description

GBE1 Monoclonal antibody specifically detects GBE1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

GBE1
OTI1D11
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
GBE1
Full length recombinant protein of human GBE1 (NP_000149) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
1,4-alpha-glucan-branching enzyme, amylo-(1,4 to 1,6) transglucosidase, amylo-(1,4 to 1,6) transglycosylase, Brancher enzyme, EC 2.4.1.18, GBE, glucan (1,4-alpha), branching enzyme 1, glycogen branching enzyme, Glycogen-branching enzyme
Mouse
IgG1
0.1 mL
Primary
2632.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit