Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GBGT1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA562371

Catalog No. PIPA562371


Add to cart

Description

Description

Immunogen sequence: HMAILADKAN GIMAAWREES HLNRHFISNK PSKVLSPEYL WDDRKPQPPS LKLIRFSTLD KDISCLR Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 46%.

Specifications

Specifications

GBGT1
Polyclonal
Unconjugated
GBGT1
Liquid
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
26301
Immunohistochemistry, Western Blot
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8N5D6
GBGT1
Recombit protein corresponding to Human GBGT1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Research Use Only