Learn More
Aviva Systems Biology Corp GBL Antibody - middle region (ARP57630_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP57630_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GBL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GBL. Alternate names: MGC111011, GBL, LST8, POP3, WAT1, GbetaLApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62916