Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syngene™ Gbox Chemi XX6
SDP

Syngene™ Gbox Chemi XX6 Ultimate offers excellent performance imaging of DNA gels, visible protein gels, chemiluminescence blots, and options for blue, green, red and IR fluorescence Westerns

Manufacturer:  Syngene™ GBOXCHEMIXX6E

 View more versions of this product

Catalog No. 01-257-168


Add to cart

Requires : PC (Not included)

Description

Description

Versatile gel documentation system for blots and gels. High performance imaging has been proven to achieve comparable sensitivity to film with the added benefit of using advanced software tools to optimise the imaging of faint bands.

Capable of many imaging applications:

  • DNA/Protein gels
  • Chemiluminescence
  • Stain free gels
  • Coloured multiplex fluorescence blots
  • 2D gels
  • In Vivo applications (additional accessories may be needed)

Peltier cooled, high resolution 6MP camera, capable of producing 16 bit images (with 65535 grey scales) with an F0.95 lens with motorized iris and focus (with feedback) for short acquisition times. Stretch darkroom has a unique motorized lift to utilise zoom features, with autofocusto ensure the maximum image resolution of the gel or blot is captured.

System comes with a seven position filter wheel, epi white light and an optional LED gantry which gives capability for fluorescence imaging, for red, green, blue and near-IR imaging (additional accessories will need to be purchased)

Features:

  • Application driven software designed for rapid image capture of biological samples
  • Simple, icon based user interface for Win10, 8 and 7 platforms
  • Takes the guess work out of imaging
  • Produce quick and simple publication quality images
  • All files produced are unique SGD format, where only raw data is analysed for quantification of samples
  • Files can be saved as TIFF, JPEG, BMP formats
  • User protocols can be saved to review previous images captured, and easily replicate future imaged gels
  • All captured images can be edited with annotations, edit tools, image sharpen and exported to GeneTools.

GeneTools software provides the user with rapid quantification of samples and goes from blots to analysed data in a few quick steps.


Analysis sample types:

  • Molecular weight
  • Colony counting
  • Spot blot
  • Band matching


All samples can be quantified, including multiplexed protein fluorescence analysis of up to 5 samples at once. Raw data files can be quickly quantified to produce true data for publication data. High quality reports can be exported in Word and Excel files for simple review.

Both GeneSys and GeneTools software meet the requirements for 21 CFR Part 11 and GLP environments.

System comes with a 3 year warranty (return to factory), unlimited licences of GeneTools analysis software and twice annual free software updates.

Specifications

Specifications

Gbox Chemi XX6 imager
115/240
7 position
4.54 x 4.54u
Windows 7 or above
Genesys and Genetools included - unlimited
optional depending on transilluminator
Gbox-Chemi-xx6
1
6mp cooled ccd
57 x 99 x 55mm
Gel / Blot Imager
50Hz
55mm
up to 25x30cm
99mm
6mp
optional
57mm
Fixed
no
removable
gels, blots, chemilumescence, fluorescence,westerns
SDS
Documents
Product Certifications
Accessories
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit