Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen GBP1
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092749
View Documents
Novus Biologicals
NBP305005100UL
100 μg Each for $320.71
Add to cart
 
NB092750
View Documents
Novus Biologicals
NBP30500520UL
20 μg Each for $138.37
Add to cart
 
Description

Description

GBP1 Polyclonal antibody specifically detects GBP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications

Specifications

GBP1
Unconjugated
Rabbit
Human, Mouse, Rat
GBP-1, GTP-binding protein 1, Guanine nucleotide-binding protein 1, guanylate binding protein 1, interferon-inducible, guanylate binding protein 1, interferon-inducible, 67kDa, huGBP-1, interferon-induced guanylate-binding protein 1
Recombinant fusion protein containing a sequence corresponding to amino acids 383-592 of human GBP1 (NP_002044.2).
Polyclonal
Affinity purified
Immunofluorescence, Immunohistochemistry, Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
2633
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit