Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GBP2 Mouse anti-Human, PE/Atto594, Clone: OTI5C8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270812PEATT594

Catalog No. N270812P594


Add to cart

Description

Description

GBP2 Monoclonal antibody specifically detects GBP2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

GBP2
OTI5C8
Flow Cytometry
Affinity Purified, Conjugated
GBP2
Full length recombinant protein of human GBP2 (NP_004111) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
GBP-2, GTP-binding protein 2, Guanine nucleotide-binding protein 2, guanylate binding protein 2, interferon-inducible, huGBP-2, interferon-induced guanylate-binding protein 2
Mouse
IgG2b
0.1 mL
Primary
2634.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit