Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GBP2 Mouse anti-Human, Clone: OTI4H2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524984

Catalog No. PIMA524984


Add to cart

Description

Description

Binds GTP, GDP and GMP. Hydrolyzes GTP very efficiently; GDP rather than GMP is the major reaction product.
Specifications

Specifications

GBP2
Monoclonal
0.52 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P32456
GBP2;Interferon-induced guanylate-binding protein 2; Interferon-induced guanylate-binding protein 2; GTP-binding protein 2; Interferon-induced guanylate-binding protein 2; Guanine nucleotide-binding protein 2
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2634
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI4H2
Unconjugated
GBP2
Liquid
GBP2
Full length human recombit protein of GBP2 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit