Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GBP5 Mouse anti-Human, Clone: OTI3G1, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA502275
Description
The guanylate-binding proteins (GBPs) are a family of interferon-induced GTP-binding proteins function in innate immunity against microbial and viral pathogens. In humans, there are seven GBPs from hGBP1 to hGBP7. Human GBP1 and GBP2 have been shown to exhibit antiviral activity as well as being able to regulate the inhibition of proliferation and invasion of endothelial cells in response to interferon (IFN). GBP5 promotes selective NALP3 inflammasome responses to pathogenic bacteria and soluble but not crystalline inflammasome priming agents.Specifications
GBP5 | |
Monoclonal | |
0.91 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96PP8 | |
GBP-5 | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
115362 |
Flow Cytometry, Western Blot | |
OTI3G1 | |
Unconjugated | |
GBP5 | |
Liquid | |
GBP5 | |
Full length human recombit protein of human GBP5 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title