Learn More
Tocris Bioscience™ GBR 12909 dihydrochloride
Selective DA uptake inhibitor; also σ ligand
Supplier: Tocris Bioscience™ 0421/10
This item is not returnable.
View return policy
Description
GBR 12909 dihydrochloride is a potent, competitive inhibitor of dopamine uptake (Ki = 1 nM for inhibition of striatal dopamine uptake). Has >100-fold lower affinity for the noradrenaline and 5-HT uptake carriers. Also a potent sigma ligand (IC50 = 48 nM). Centrally active following systemic administration.Specifications
1-[2-[Bis-(4-fluorophenyl)methoxy]ethyl]-4-(3-phenylpropyl)piperazine dihydrochloride | |
>98% | |
Soluble to 50mM in DMSO | |
Observed MW: 523.49 |
67469-78-7 | |
10 mg | |
C28H32F2N2O.2HCl |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature