Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GBX2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540441
Description
Peptide sequence: QGKDESKVED DPKGKEESFS LESDVDYSSD DNLTGQAAHK EEDPGHALEE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 85%; Pig: 100%; Rabbit: 100%; Rat: 100%.
Altered expression of GBX2, part of the homeobox-containing human family of DNA-binding transcription factors, is associated with therapy failure and death in patients with multiple types of cancer.
Specifications
GBX2 | |
Polyclonal | |
Unconjugated | |
GBX2 | |
D130058E05Rik; gastrulation and brain-specific homeobox protein 2; gastrulation brain homeobox 2; Gbx2; Gbx-2; Homeobox protein GBX-2; LOW QUALITY PROTEIN: homeobox protein GBX-2; MMoxA; Stimulated by retinoic acid gene 7 protein; Stra7 | |
Rabbit | |
Protein A | |
RUO | |
2637 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P52951 | |
GBX2 | |
Synthetic peptide directed towards the middle region of human GBX2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction