Learn More
Trajan™ BP624 GC Capillary Columns
SGE BP624 GC Capillary Columns - A polar phase designed for EPA volatile organics analysis
Manufacturer:  Trajan™ 054832
 View more versions of this product
Catalog No. 03-350-368
Add to cart
Description
Description
  • Polar phase
  • EPA organics analysis (EPA 624, 502.2, SW-846, 8240/8260)
  • Ideal for environmental analyses, volatile organics, alcohol analysis
Specifications
Specifications
0.32 mm
230/240°C
1.8 µm
30 m