Learn More
Trajan™ BP624 GC Capillary Columns
SGE BP624 GC Capillary Columns - A polar phase designed for EPA volatile organics analysis
$691.50 - $1193.00
Specifications
Max. Temperature 230/240°C
View More Specs
Products 3
Catalog Number Mfr. No. Diameter (Metric) Inner Film Thickness Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Film Thickness Length (Metric) Price Quantity & Availability  
03-350-369
View Documents
Trajan™
054836
0.53 mm 3 mm 30 mm
Each for $691.50
Add to cart
03-350-368
View Documents
Trajan™
054832
0.32 mm 1.8 µm 30 m
Each for $698.00
Add to cart
SG-054835
View Documents
Trajan™
054835
0.53 mm 3 µm 50 m
Each for $1,193.00
Add to cart
Description
Description
  • Polar phase
  • EPA organics analysis (EPA 624, 502.2, SW-846, 8240/8260)
  • Ideal for environmental analyses, volatile organics, alcohol analysis
Specifications
Specifications
230/240°C