Learn More
GC Mouse anti-Human, Clone: OTI2A1, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809264
Catalog No. 50-166-9380
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
GC (vitamin D-binding protein) is involved in vitamin D transport/storage, scavenging of extracellular G-actin, enhancement of the chemotactic activity of C5 alpha for neutrophils in inflammation and macrophage activation.
Specifications
Specifications
GC
Monoclonal
Unconjugated
GC
Lyophilized
GC
Full length human recombit protein of human GC produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI2A1
PBS with 8% trehalose and no preservative; pH 7.3
P02774
DBP, DBP-maf, DBP/GC, GRD3, Gc-MAF, GcMAF, HEL-S-51, VDB, VDBG, VDBP
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
2638