Learn More
Thermo Scientific™ GC-Orbitrap™ High Temperature Nut and Ferrule Kit
Allows for leak-free, high temperature transferline operation on Orbitrap-GC/MS with the ease-of-use of Vespel ferrules.
$234.50 - $468.50
Products 3
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
03-251-524
View Documents
Thermo Scientific™
1R1204340020
Replacement GC-Orbitrap Spring-Loaded Nut Each for $234.50
Add to cart
03-251-523
View Documents
Thermo Scientific™
290VT221
High Temperature Ferrule Replacement Pk. Pack of 5 for $234.50
Add to cart
03-251-530
View Documents
Thermo Scientific™
1R1204340100
GC-Orbitrap High Temperature Nut & Ferrule Kit Each for $468.50
Add to cart
Description
Description
Use the Thermo Scientific™ GC-Orbitrap™ High Temperature Nut and Ferrule Kit to obtain leak-free results at high temperature transferline settings on your gas chromatography (GC) Orbitrap. The high-temperature SCP-5000 Vespel ferrules (included) allow continuous operating transferline temperatures of at least 320°C without ferrule deformation or breakdown (common with standard graphite/Vespel ferrules above 280°C), preventing leaks from forming over an extended period of time. The included spring loaded nut ensures a close fit between nut, ferrule and transferline, further preventing leaks from forming.
High-temperature SCP-5000 Vespel ferrules with 0.4mm ID can be used with most 0.1–0.25mm ID gas chromatography columns. Ferrules must be used in conjunction with High Temperature Adjustable Transferline Nut for GC-Orbitrap (included in kit). Nut and ferrules are for use with the Thermo Scientific™ Q Exactive™ GC Orbitrap™ GC-MS/MS system and the Thermo Scientific™ Exactive™ GC Orbitrap™ GC-MS system.