Learn More
Trajan Scientific™ Shimadzu Plug Style Premium High-Temperature HT Septa
SDP
Retains softness and pierceability at high temperatures and low injection port adhesion.
Supplier:  Trajan Scientific™ 041895
Catalog No. 04-189-5
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. View substitute products »
Description
Description
  • Bleed and temperature optimized
  • Reliable seal against carrier gas pressure in the inlet
  • No bleed or contamination into the GC system
  • Maximum operating temperature of 400°C (330°C for 17mm size)
Specifications
Specifications
BTO Silicone
400°C
HT GC Plug
50/Pk.