Learn More
MilliporeSigma™ Supelco™ GC Stationary Phase, SF-96
A methyl silicone stationary phase suitable for gas chromatography
Manufacturer:  MilliporeSigma™ Supelco™ 21101U
Catalog No. 11-100-2521
Add to cart
Description
Description
  • Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase
Specifications
Specifications
250°C
SF-96
Bottle
50 g