Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCAP2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP17977420UL

 View more versions of this product

Catalog No. NBP17977420


Add to cart

Description

Description

GCAP2 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

GCAP2
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
GUCA1B
Synthetic peptide directed towards the N terminal of human GUCA1B. Peptide sequence SGTLFMHEFKRFFKVTDDEEASQYVEGMFRAFDKNGDNTIDFLEYVAALN.
23 kDa
20ul
Signal Transduction, Vision
Primary
2979
Western Blot
Western Blot 1:1000
NP_002089
DKFZp686E1183, GCAP 2, GCAP2guanylate cyclase-activating protein, photoreceptor 2, Guanylate cyclase activator 1B, guanylate cyclase activator 1B (retina), guanylyl cyclase-activating protein 2, GUCA2, RP48
Rabbit
IgG
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit