Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCH1 Monoclonal Antibody (OTI7B8)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527278

Catalog No. PIMA527278


Only null left
Add to Cart

Description

Description

GCH1 Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the GTP cyclohydrolase family. The encoded protein is the first and rate-limiting enzyme in tetrahydrobiopterin biosynthesis, catalyzing the conversion of GTP into 7,8-dihydroneopterin triphosphate. BH4 is an essential cofactor required by aromatic amino acid hydroxylases as well as nitric oxide synthases. Mutations in this gene are associated with malignant hyperphenylalaninemia and dopa-responsive dystonia. Several alternatively spliced transcript variants encoding different isoforms have been described; however, not all variants give rise to a functional enzyme.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GCH1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P30793
GCH1
Full length human recombinant protein of GCH1 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
OTI7B8
Unconjugated
GCH1
dystonia 14; DYT14; DYT5; DYT5a; GCH; Gch1; GTP cyclohydrolase 1; GTP cyclohydrolase I; GTPCH; GTP-CH; GTPCH1; GTP-CH-1; GTP-CH-I; guanosine 5'-triphosphate cyclohydrolase I; HPABH4B
Mouse
Affinity Chromatography
RUO
2643
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.