Learn More
GCH1 Mouse anti-Human, Clone: OTI7B8, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF810250
Catalog No. 50-166-9971
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the GTP cyclohydrolase family. The encoded protein is the first and rate-limiting enzyme in tetrahydrobiopterin biosynthesis, catalyzing the conversion of GTP into 7,8-dihydroneopterin triphosphate. BH4 is an essential cofactor required by aromatic amino acid hydroxylases as well as nitric oxide synthases. Mutations in this gene are associated with maligt hyperphenylalaninemia and dopa-responsive dystonia. Several alternatively spliced transcript variants encoding different isoforms have been described; however, not all variants give rise to a functional enzyme.
Specifications
Specifications
GCH1
Monoclonal
Unconjugated
GCH1
Lyophilized
GCH1
Full length human recombit protein of human GCH1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI7B8
PBS with 8% trehalose and no preservative; pH 7.3
P30793
DYT14, DYT5, DYT5a, GCH, GTP-CH-1, GTPCH1, HPABH4B
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
2643