Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCHFR Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549121

Catalog No. PA549121

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine based on sequence homology.

Mediates tetrahydrobiopterin inhibition of GTP cyclohydrolase 1. This inhibition is reversed by L-phenylalanine.
Specifications

Specifications

GCHFR
Polyclonal
PBS with 0.09% sodium azide
P30047
GTP cyclohydrolase 1 feedback regulatory protein, GFRP, GTP cyclohydrolase I feedback regulatory protein, p35, GCHFR, GFRP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2644
Western Blot
Unconjugated
GCHFR
Liquid
GCHFR
KLH conjugated synthetic peptide between 38-66 amino acids from the C-terminal region of human GCHFR.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit