Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCKR Antibody (OTI1E7), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270823JF646
Description
GCKR Monoclonal specifically detects GCKR in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
GCKR | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GCKR | |
Full length recombinant protein of human GCKR (NP_001417) produced in HEK293T cells. | |
0.1 mL | |
2646.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1E7 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
FGQTL5, GKRP, glucokinase (hexokinase 4) regulator, glucokinase (hexokinase 4) regulatory protein, Glucokinase regulator, glucokinase regulatory protein | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction