Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GCKR Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115275
Description
Antibody detects endogenous levels of total GCKR.
This gene encodes a protein belonging to the GCKR subfamily of the SIS (Sugar ISomerase) family of proteins. The gene product is a regulatory protein that inhibits glucokinase in liver and pancreatic islet cells by binding non-covalently to form an inactive complex with the enzyme. This gene is considered a susceptibility gene candidate for a form of maturity-onset diabetes of the young (MODY).
Specifications
GCKR | |
Polyclonal | |
Unconjugated | |
Gckr | |
FGQTL5; Gckr; GKRP; GLRE; glucokinase (hexokinase 4) regulator; glucokinase regulator; Glucokinase regulatory protein; hexokinase 4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
231103, 25658, 2646 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q07071, Q14397, Q91X44 | |
Gckr | |
A synthesized peptide derived from human GCKR(Accession Q14397), corresponding to amino acid residues G207-S257. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction