Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCLC Mouse anti-Human, Rat, Monkey, Alexa Fluor™ 647, Clone: OTI1A3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270830AF647

Catalog No. N270830A647


Add to cart

Description

Description

GCLC Monoclonal antibody specifically detects GCLC in Human, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

GCLC
OTI1A3
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
GCLC
Full length recombinant protein of human GCLC (NP_001489) produced in HEK293T cells.
Immunogen affinity purified
Cell Biology, Lipid and Metabolism
Primary
2729.0
Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
EC 6.3.2.2, gamma-ECS, Gamma-glutamylcysteine synthetase, GCS, GCS heavy chain, GLCL, GLCLCGCL, glutamate--cysteine ligase catalytic subunit, glutamate-cysteine ligase, catalytic subunit
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Monkey, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit