Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCLC Antibody (OTI1A3), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270830AF700
Description
GCLC Monoclonal specifically detects GCLC in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
GCLC | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GCLC | |
Full length recombinant protein of human GCLC (NP_001489) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Rat, Monkey | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1A3 | |
Western Blot, Immunohistochemistry | |
EC 6.3.2.2, gamma-ECS, Gamma-glutamylcysteine synthetase, GCS, GCS heavy chain, GLCL, GLCLCGCL, glutamate--cysteine ligase catalytic subunit, glutamate-cysteine ligase, catalytic subunit | |
Mouse | |
Affinity Purified | |
Cell Biology, Lipid and Metabolism | |
2729.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction