Learn More
Aviva Systems Biology Corp GCM1 Antibody - C-terminal region (P100837_P050) | 100uL
Supplier:  Aviva Systems Biology Corp P100837_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GCM1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GCM1. Alternate names: GCMA, hGCMaApplicationWB. Species reactivity: Dog, Horse, Human, Rabbit
Catalog No. 50-014-62737