Learn More
GCM1 Mouse anti-Human, Clone: OTI1A4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810523

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a DNA-binding protein with a gcm-motif (glial cell missing motif). The encoded protein is a homolog of the Drosophila glial cells missing gene (gcm). This protein binds to the GCM-motif (A/G)CCCGCAT, a novel sequence among known targets of DNA-binding proteins. The N-terminal DNA-binding domain confers the unique DNA-binding activity of this protein.Specifications
GCM1 | |
Monoclonal | |
Unconjugated | |
GCM1 | |
GCMA, hGCMa | |
Mouse | |
Affinity Chromatography | |
RUO | |
8521 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1A4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NP62 | |
GCM1 | |
Full length human recombit protein of human GCM1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.