Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCNF/NR6A1 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI6F3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270825AF405

Catalog No. N270825A405


Add to cart

Description

Description

GCNF/NR6A1 Monoclonal antibody specifically detects GCNF/NR6A1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

GCNF/NR6A1
OTI6F3
Western Blot
Affinity Purified, Conjugated
NR6A1
Recombinant protein fragment corresponding to amino acids 1-248 of human GCNF/NR6A1 (NP_001480) produced in E.coli.
Immunogen affinity purified
GPCR, Olfactory
Primary
2649.0
Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
GCNF1, GCNFNR61, Germ cell nuclear factor, hGCNF, nuclear receptor subfamily 6 group A member 1, nuclear receptor subfamily 6, group A, member 1, retinoic acid receptor-related testis-associated receptor, Retinoid receptor-related testis-specific receptor, RTRhRTR
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit