Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCOM1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Specifications
Antigen | GCOM1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP155528
![]() |
Novus Biologicals
NBP155528 |
100 μL | N/A | N/A | N/A | ||||
Description
GCOM1 Polyclonal specifically detects GCOM1 in Human samples. It is validated for Western Blot.Specifications
GCOM1 | |
Polyclonal | |
Rabbit | |
Q6EES4 | |
145781 | |
Synthetic peptides corresponding to GCOM1 The peptide sequence was selected from the C terminal of GCOM1. Peptide sequence ERMEKERHQLQLQLLEHETEMSGELTDSDKERYQQLEEASASLRERIRHL. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Gcom, GRINL1A combined protein, GRINL1A upstream protein, Gup | |
GCOM1 | |
IgG | |
52 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title