Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCP3/Gamma-tubulin complex component 3 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS13277R

Description
The gamma-Tubulin complex is composed of gamma Tubulin and the gamma-Tubulin complex associated proteins GCP2, GCP3, GCP4, GCP5 and GCP6, all of which are essential components of microtubule organizing centers. gamma-Tubulin complex components are localized to both the centrosome, where they are involved in microtubule nucleation, and to the cytoplasm, where they exist as soluble complexes that can be recruited to the centrosome as needed. Although the GCP proteins are related, they have distinct roles which contribute to the proper function of the gamma-Tubulin complex. GCP3 (gamma-tubulin complex component 3), also known as TUBGCP3 or SPBC98, localizes to the centrosome and is an ubiquitously expressed 907 amino acid member of the gamma-Tubulin complex.Specifications
GCP3/Gamma-tubulin complex component 3 | |
Polyclonal | |
Unconjugated | |
TUBGCP3 | |
104p; Gamma-ring complex protein 104 kDa; Gamma-tubulin complex component 3; GCP3; GCP-3; Grip104; h104p; hGCP3; hGrip104; hSpc98; SPBC98; Spc98; Spc98p; spindle pole body protein; Spindle pole body protein Spc98 homolog; Tubgcp3; tubulin gamma complex associated protein 3; tubulin, gamma complex associated protein 3 | |
Rabbit | |
Protein A | |
RUO | |
10426 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q96CW5 | |
TUBGCP3 | |
KLH conjugated synthetic peptide derived from human GCP3/Gamma-tubulin complex component 3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction