Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCP4 Mouse anti-Human, DyLight 680, Clone: 812, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243586FR

Catalog No. NBP243586FR


Add to cart

Description

Description

GCP4 Monoclonal antibody specifically detects GCP4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

GCP4
812
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
TUBGCP4
Recombinant protein encompassing a sequence within the center region of human GCP4. The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
76PhGCP4, FLJ14797, gamma tubulin ring complex protein (76p gene), Gamma-ring complex protein 76 kDa, gamma-tubulin complex component 4, GCP-4, GCP4hGrip76, h76p, tubulin, gamma complex associated protein 4
Mouse
IgG2b
0.1 ml
Primary
27229
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit