Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCSH Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548809

Catalog No. PA548809

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine and rabbit based on sequence homology.

The glycine cleavage system catalyzes the degradation of glycine. The H protein (GCSH) shuttles the methylamine group of glycine from the P protein (GLDC) to the T protein (GCST).
Specifications

Specifications

GCSH
Polyclonal
PBS with 0.09% sodium azide
P23434
Glycine cleavage system H protein, mitochondrial, Lipoic acid-containing protein, GCSH
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2653
Western Blot
Unconjugated
GCSH
Liquid
GCSH
KLH conjugated synthetic peptide between 27-56 amino acids from the N-terminal region of human GCSH.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit