Learn More
GD2 Mouse anti-Human, BUV395, Clone: 14.G2a, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565990
Catalog No. BDB565990
Add to cart
Description
Description
GD2 Monoclonal antibody specifically detects GD2 in Human samples. It is validated for Flow cytometry.
Specifications
Specifications
GD2
14.G2a
Vol. Per test: 5μL
Purified
Mouse
Mouse BALB/c IgG2a
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BUV395 under optimum conditions, and unconjugated antibody and free BD Horizon™ BUV395 were removed.
Monoclonal
Flow Cytometry
Brilliant Ultraviolet 395
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
GD2
LAN-1 human neuroblastoma cells
25 Tests
Cell Biology
Primary
Human
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.