Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GDA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530322
Description
Recommended positive controls: HeLa, HCT116. Predicted reactivity: Mouse (88%), Rat (88%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GDA is an enzyme that catalyzes the hydrolytic deamination of guanine, producing xanthine and ammonia.
Specifications
GDA | |
Polyclonal | |
Unconjugated | |
GDA | |
AU015411; AW047581; Cypin; cytoplasmic PSD95 interactor; GAH; Gda; GUANASE; guanine aminase; guanine aminohydrolase; Guanine deaminase; KIAA1258; NEDASIN; p51-nedasin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14544, 83585, 9615 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.84 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9R111, Q9WTT6, Q9Y2T3 | |
GDA | |
Recombinant fragment corresponding to a region within amino acids 52 and 359 of Human Guanine deaminase. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction