Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GDA Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | GDA |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119167
|
Invitrogen™
PA5119167 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121962
|
Invitrogen™
PA5121962 |
20 μL |
Each for $209.50
|
|
|||||
Description
GDA Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
GDA is an enzyme that catalyzes the hydrolytic deamination of guanine, producing xanthine and ammonia.Specifications
GDA | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
GDA | |
AU015411; AW047581; Cypin; cytoplasmic PSD95 interactor; GAH; Gda; GUANASE; guanine aminase; guanine aminohydrolase; Guanine deaminase; KIAA1258; NEDASIN; p51-nedasin | |
GDA | |
Primary | |
Antigen affinity chromatography |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9Y2T3 | |
9615 | |
Recombinant protein Guanine Deaminase. The antigen corresponds to amino acid range 1-454 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title