Learn More
Azzota Corp GDC-0032 DFS Item
Manufacturer:  Azzota Corp C1139
GDC-0032 (Taselisib) is a selective, potent, orally bioavailable inhibitor of PI3Ka with a Ki = 0.2nM, and with reduced inhibitory activity against PI3Kβ. Please request if need more quantities
Catalog No. 50-161-3106