Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDE1 Mouse anti-Human, Clone: 2021CT823.61.7, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Thermo Scientific MA537775

Encompass

Catalog No. PIMA537775


Add to cart

Description

Description

GDE1 Monoclonal antibody specifically detects GDE1 in Human samples. It is validated for Western Blot
Specifications

Specifications

GDE1
Monoclonal
Lot-Specific
PBS with 0.09% sodium azide
Q9NZC3
1200003M13Rik; 363E6.2; Gde1; glycerophosphodiester phosphodiesterase 1; membrane interacting protein of RGS16; membrane-interacting protein of RGS16; MIR16; PAMCI; PCIP1; RGS16; RGS16-interacting membrane protein
recombinant protein from the human region of human GDE1
Protein G
RUO
Antibody
Monoclonal
Human
Western Blot
2021CT823.61.7
Unconjugated
Gde1
Liquid
Mouse
IgG1, kappa
200 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
51573
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit