Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDEP Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 227441AP

Catalog No. 50-173-3938


Add to cart

Specifications

Specifications

GDEP
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Liquid
PCAT4
GDEP Fusion Protein Ag19087
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PCAT4
GDEP, PCAN1
Rabbit
IgG
150 μL
-20°C
Primary
118425
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit