Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ GDF-11/BMP-11 Human Recombinant Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

Supplier:  Novus Biologicals™ NBP2520300.1MG

 View more versions of this product

Catalog No. NBP25203001


Add to cart

Description

Description

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 299-407 of Human GDF-11/BMP-11 The Recombinant Human GDF-11/BMP-11 Protein is derived from E. coli. The Recombinant Human GDF-11/BMP-11 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

1mg/mL
20mM Tris-HCl (pH8.0) containing 10% glycerol
TMW: 14.8kDa
0.1mg
BMP-11BMP11Bone morphogenetic protein 11, GDF-11, growth differentiation factor 11, growth/differentiation factor 11
Unlabeled
SDS-Page
10220
>85%, by SDS-PAGE
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
GDF-11/BMP-11
Yes
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For research use only and is not approved for use in humans or in clinical diagnosis