Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDF-15 Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0307-10A3, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212331AF350
Description
GDF-15 Monoclonal specifically detects GDF-15 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
GDF-15 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
GDF15 | |
Human recombinant GDF-15 | |
Primary | |
This antibody was selected for its ability to detect human GDF-15. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
MM0307-10A3 | |
Western Blot, Flow Cytometry | |
growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDFGDF-15, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGFBPTGF-beta | |
Mouse | |
0.1 mL | |
9518 | |
Human | |
IgG2 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title