Learn More
GDF-15 Mouse anti-Human, Alexa Fluor 488, Clone: MM0307-10A3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212331AF488
Catalog No. NB212331488
Add to cart
Description
Description
GDF-15 Monoclonal specifically detects GDF-15 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
Specifications
GDF-15
MM0307-10A3
Western Blot, Flow Cytometry
Purified
GDF15
Human GDF15
Protein G purified
Hepatic Stem Cell Markers, Stem Cell Markers
Primary
9518
Human
Western Blot, Flow Cytometry
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDFGDF-15, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGFBPTGF-beta
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody was selected for its ability to detect human GDF-15.