Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDF-15 Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0307-10A3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212331AF594

Catalog No. NP212331A94


Add to cart

Description

Description

GDF-15 Monoclonal antibody specifically detects GDF-15 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

GDF-15
MM0307-10A3
Western Blot, Flow Cytometry
Conjugated, Purified
GDF15
Human GDF15
Protein G purified
Hepatic Stem Cell Markers, Stem Cell Markers
Primary
9518.0
Human
Flow Cytometry, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDFGDF-15, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGFBPTGF-beta
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
This antibody was selected for its ability to detect human GDF-15.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit