Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDF-8/Myostatin Antibody (RM0243-13F2), Alexa Fluor™ 750, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212334AF750
Description
GDF-8/Myostatin Monoclonal specifically detects GDF-8/Myostatin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GDF-8/Myostatin | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
MSTN | |
Mouse recombinant GDF-8/Myostatin | |
0.1 mL | |
Primary | |
Mouse | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0243-13F2 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, Myostatin | |
Rat | |
Protein A or G purified | |
Cytokine Research | |
2660.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction