Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ GDF-8/Myostatin Human Recombinant Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

Manufacturer:  Novus Biologicals™ NBP2519240.02MG

 View more versions of this product

Catalog No. NBP25924002


Add to cart

Description

Description

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 267-375 of Human GDF-8/Myostatin The Recombinant Human GDF-8/Myostatin Protein is derived from E. coli. The Recombinant Human GDF-8/Myostatin Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

0.25mg/mL
20mM Tris-HCl (pH8.0) containing 10% glycerol
TMW: 15.1kDa
0.02mg
GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, Myostatin
Unlabeled
SDS-Page
2660
>90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
GDF-8/Myostatin
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For research use only and is not approved for use in humans or in clinical diagnosis