Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDF-8/Myostatin Mouse anti-Human, Clone: GDF9/4261, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen GDF-8/Myostatin
Clone GDF9/4261
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, ELISA
Applications Western Blot, ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122885
View Documents
Novus Biologicals
NBP30770610UG
100 μg
Each for $352.89
Add to cart
 
NB122886
View Documents
Novus Biologicals
NBP30770620UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

GDF-8/Myostatin Monoclonal antibody specifically detects GDF-8/Myostatin in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

GDF-8/Myostatin
0.2 mg/ml
Western Blot, ELISA
Purified
RUO
Human
GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, Myostatin
Recognizes the 113 amino acid carboxy-terminal fragment of GDF-8/Myostatin protein
Monoclonal
Protein A or G purified
GDF9/4261
Western Blot 1-2 ug/ml, ELISA
Unconjugated
Mouse
Cytokine Research
10 mM PBS with 0.05% BSA
2660
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit