Learn More
GDF-8/Myostatin Mouse anti-Human, DyLight 405, Clone: 6E4E6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237264V
Catalog No. NBP237264V
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
GDF-8/Myostatin
6E4E6
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
MSTN
Purified recombinant fragment of human MSTN (AA:24-266) expressed in E. Coli.
Protein A or G purified
Cytokine Research
Primary
2660
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, Myostatin
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human