Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDF-8/Myostatin Mouse anti-Human, FITC, Clone: 6E4E6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237264F

Catalog No. NBP237264F

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

GDF-8/Myostatin
6E4E6
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
MSTN
Purified recombinant fragment of human MSTN (AA:24-266) expressed in E. Coli.
Protein A or G purified
Cytokine Research
Primary
2660
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
FITC
PBS with 0.05% Sodium Azide
GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, Myostatin
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit