Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDF-8/Myostatin Rat anti-Mouse, Alexa Fluor™ 350, Clone: RM0243-13F2, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212334AF350

Catalog No. NP212334A30


Add to cart

Description

Description

GDF-8/Myostatin Monoclonal antibody specifically detects GDF-8/Myostatin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

GDF-8/Myostatin
RM0243-13F2
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
MSTN
Mouse GDF8/Myostatin
Protein G purified
Cytokine Research
Primary
2660.0
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, Myostatin
Rat
IgG2
0.1 mL
Store at 4°C in the dark.
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit