Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDF-9 Rat anti-Human, DyLight 405, Clone: 72B, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP261933V
Description
GDF-9 Monoclonal antibody specifically detects GDF-9 in Human samples. It is validated for Western Blot, ELISA.Specifications
GDF-9 | |
72B | |
Western Blot, ELISA | |
Purified | |
GDF9 | |
N-terminal of GDF-9 corresponding to sequence KKPLGPASFNLSEYFC | |
Protein G purified | |
Cytokine Research | |
Primary | |
2661 |
ELISA, Western Blot | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GDF9, GDF-9, growth differentiation factor 9, growth/differentiation factor 9 | |
Rat | |
IgG1 | |
0.1mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title